missing translation for 'onlineSavingsMsg'
Learn More

Mouse Sulfamidase/SGSH Antibody, R&D Systems™

Código de producto. 18000882
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18000882 100 μg 100 microgramos
18784957 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18000882 Proveedor R&D Systems N.º de proveedor AF2969

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

Sulfamidase/SGSH Polyclonal specifically detects Sulfamidase/SGSH in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Sulfamidase/SGSH
Aplicaciones Western Blot, Immunoprecipitation
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
N.º de referencia del gen NP_061292
Alias de gen heparan sulfate sulfatase, HSS, HSSN-sulphoglucosamine sulphohydrolase, MPS3A, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMD, SFMDEC 3.10.1.1, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase
Símbolos de los genes SGSH
Especie del huésped Goat
Inmunógeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Sulfamidase/SGSH Arg23-Leu502 Accession # NP_061292
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6448
Especificidad de la prueba Detects mouse Sulfamidase/SGSH in direct ELISAs and Western blots.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.