Learn More
R&D Systems™ Mouse STING/TMEM173 (NP_082537) VersaClone cDNA
Descripción
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Información de órdenes
Ships at ambient temperature
Especificaciones
Especificaciones
| génica | STING/TMEM173 |
| Número de acceso | NP_082537 |
| Concentración | 10 μg at 0.2 μg/μL |
| Formulación | cDNA is provided in 10mM Tris-Cl, pH 8.142 |
| Tipo de inserto | 1150bp |
| ID de gen (Entrez) | 340061 |
| Alias de gen | endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Mouse |
| Cantidad | 10 μg |
For Reasearch Use Only. Not for use in Humans.