Learn More
Mouse/Rat RAGE Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18705568
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18705568 25 μg 25 microgramos
18754387 100 μg 100 microgramos
18704368 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18705568 Proveedor R&D Systems N.º de proveedor MAB1179SP
Rat Monoclonal Antibody has been used in 15 publications
AGER Monoclonal specifically detects AGER in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RAGE
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon 175410
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen NP_031451
Alias de gen advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Símbolos de los genes Ager
Especie del huésped Rat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Ala342 Accession # NP_031451
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5891
Especificidad de la prueba Detects mouse and rat RAGE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RAGE is observed.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Mouse, Rat
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2a