missing translation for 'onlineSavingsMsg'
Learn More

Mouse Latexin Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18737547
Cambiar vista
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18737547 25 μg 25 microgramos
18070872 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18737547 Proveedor R&D Systems N.º de proveedor AF3620SP

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Sheep Polyclonal Antibody

Latexin Polyclonal specifically detects Latexin in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Latexin
Aplicaciones Western Blot, Immunoprecipitation, Neutralization
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.4 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P70202
Alias de gen ECI, ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, LXN, TCI, TCIProtein MUM, Tissue carboxypeptidase inhibitor
Símbolos de los genes LXN
Especie del huésped Sheep
Inmunógeno E. coli-derived recombinant mouse Latexin Glu2-Glu222 Accession # P70202
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 56925
Especificidad de la prueba Detects mouse Latexin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human Latexin is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.