Learn More
R&D Systems™ Mouse Desmoglein-2 (NP_031909.2) VersaClone cDNA
Especificaciones
Especificaciones
| génica | Desmocollin-2 |
| Número de acceso | NP_031909.2 |
| ID de gen (Entrez) | 1824 |
| Alias de gen | ARVD11, Cadherin family member 2, CDHF2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC2, DSC3DKFZp686I11137 |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Mouse |
| Cantidad | 10 μg |