Learn More
R&D Systems™ Mouse Coagulation Factor II (NP_034298.1) VersaClone cDNA
Especificaciones
Especificaciones
| génica | Coagulation Factor II/Thrombin |
| Número de acceso | NP_034298.1 |
| ID de gen (Entrez) | 2147 |
| Alias de gen | Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), F2, PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin, Thrombin receptor-like 2 |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Mouse |
| Cantidad | 10 μg |