missing translation for 'onlineSavingsMsg'
Learn More

ZAP70, Mouse, Clone: ZAP-03, Abnova™

Código de producto. 16055906
Cambiar vista
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16055906 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16055906 Proveedor Abnova N.º de proveedor MAB3664.100ug

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse monoclonal antibody raised against partial recombinant ZAP70.

This gene encodes an enzyme belonging to the protein tyrosine kinase family, and it plays a role in T-cell development and lymphocyte activation. This enzyme, which is phosphorylated on tyrosine residues upon T-cell antigen receptor (TCR) stimulation, functions in the initial step of TCR-mediated signal transduction in combination with the Src family kinases, Lck and Fyn. This enzyme is also essential for thymocyte development. Mutations in this gene cause selective T-cell defect, a severe combined immunodeficiency disease characterized by a selective absence of CD8-positive T-cells. Two transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq

Especificaciones

Antígeno ZAP70
Aplicaciones Flow Cytometry, Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon ZAP-03
Conjugado Unconjugated
Descripción Mouse monoclonal antibody raised against partial recombinant ZAP70.
Dilución Flow Cytometry (2-5 ug/mL) Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In PBS, pH 7.4 (0.09% sodium azide)
génica ZAP70
Alias de gen FLJ17670/FLJ17679/SRK/STD/TZK/ZAP-70
Símbolos de los genes ZAP70
Especie del huésped Mouse
Inmunógeno Recombinant His fusion protein corresponding to 160 amino acids of human ZAP70.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7535
Especificidad de la prueba This antibody reacts with ZAP70, a 70 KDa protein tyrosine kinase expressed in T and NK cells. ZAP70 is a molecule susceptible to degradation. It is recommended to use freshly prepared cell lysates (protease inhibitors are essential) to avoid non-specific staining of degradation products.
Especies diana Human
Contenido y almacenamiento Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.