Learn More
PHB, Mouse, Clone: 5H7, Abnova™
Descripción
Prohibitin is an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | PHB |
| Aplicaciones | ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Clasificación | Monoclonal |
| Clon | 5H7 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against recombinant PHB. |
| Dilución | ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
| Formulación | In ascites (0.03% sodium azide) |
| génica | PHB |
| Alias de gen | PHB1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.