Learn More
MMP3, Mouse, Clone: SB14d, Abnova™
Descripción
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.[provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | MMP3 |
| Aplicaciones | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Clasificación | Monoclonal |
| Clon | SB14d |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against recombinant MMP3. |
| Dilución | ELISA (1:3000-1:8000) Immunohistochemistry (2 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulación | In 100mM BBS, pH 8.2 |
| génica | MMP3 |
| Alias de gen | CHDS6/MGC126102/MGC126103/MGC126104/MMP-3/SL-1/STMY/STMY1/STR1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.