Learn More
Invitrogen™ Mouse anti-Human IgE Secondary Antibody
Código de producto. 11505030
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
11505030 100 μg 100 microgramos
1 options
Código de producto. 11505030 Proveedor Invitrogen™ N.º de proveedor SA119259
Mouse Monoclonal Secondary Antibody
This antibody recognizes an epitope different from the ones recognized by 4G7 and 4H10 antibodies to IgE.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Human IgE
Aplicaciones ELISA, Flow Cytometry
Clasificación Monoclonal
Clon BE5
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 15mM sodium azide; pH 7.4
Especie del huésped Mouse
Inmunógeno Purified human IgE.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG1