Learn More
COX17, Mouse, Polyclonal Antibody, Abnova™
Descripción
Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex. This nuclear gene encodes a protein which is not a structural subunit, but may be involved in the recruitment of copper to mitochondria for incorporation into the COX apoenzyme. This protein shares 92% amino acid sequence identity with mouse and rat Cox17 proteins. This gene is no longer considered to be a candidate gene for COX deficiency. A pseudogene COX17P has been found on chromosome 13. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | COX17 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse polyclonal antibody raised against a partial recombinant COX17. |
| Formulación | 50% glycerol |
| génica | COX17 |
| N.º de referencia del gen | NM_005694 |
| Alias de gen | MGC104397/MGC117386 |
| Símbolos de los genes | COX17 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.