missing translation for 'onlineSavingsMsg'
Learn More

CD326 Mouse anti-Human, PE-CF594, Clone: EBA-1, BD

Código de producto. 15804299
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15804299 50 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15804299 Proveedor BD Biosciences N.º de proveedor 565399

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The EBA-1 monoclonal antibody specifically binds to human CD326. CD326 is an approximately 40 kDa type 1 transmembrane glycoprotein and adhesion molecule that mediates intercellular adhesive interactions. CD326 is also known as epithelial adhesion molecule (EpCAM), epithelial glycoprotein 2 (EGP-2), and epithelial surface antigen (ESA). The epithelial cells present in non-squamous epithelia and tumors derived from such cells show EpCAM expression. The normal epithelial cells reactive with anti-EpCAM antibodies are those present in the (lower) respiratory tract; the (lower) gastrointestinal tract; tubules in the kidney; the surface epithelium of the ovary; the exocrine and endocrine pancreas; secondary germ cells of telogenic hair follicles; and secretory tubules of sweat glands in the skin, whereas the epidermis is negative. In addition, all epithelial cells in the thyroid and epithelial cells in the thymus show EpCAM expression, while the outer cortex and Hassall’s corpuscles have low expression. In the liver, only the bile ducts appear to be positive with anti-EpCAM antibodies. Non-squamous- carcinoma cells have high EpCAM expression; some squamous carcinoma cells. Tumors arising from non-epithelial cells, such as lymphoma, mesothelioma, neuroblastoma, and melanoma, do not express EpCAM.

This antibody is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg, 610/20-nm filter).

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD326
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon EBA-1
Conjugado PE-CF594
Descripción EPCAM; EGP; ESA; GA733-2; hEGP-2; KSA; M4S1; MIC18; MK-1; TACSTD1; TROP1
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Breast carcinoma-associated mucin BCA-225
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 λ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.