Learn More
Motesanib, MedChemExpress
Código de producto. 16480107
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16480107 50 mg 50 miligramos
16454796 100 mg 100 miligramos
16423728 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16480107 Proveedor Medchem Express N.º de proveedor HY10228
MedChemExpress Motesanib (AMG 706) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50s of 2 nM/3 nM/6 nM, respectively, and has similar activity against Kit, and is appr 10-fold more selective for VEGFR than PDGFR and Ret.
Motesanib (AMG 706) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50s of 2 nM/3 nM/6 nM, respectively, and has similar activity against Kit, and is appr 10-fold more selective for VEGFR than PDGFR and Ret.
Especificaciones
Nombre del producto químico o material Motesanib
CAS 453562-69-1
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C22H23N5O
Cantidad 50 mg
Sinónimo AMG 706
Información de solubilidad DMSO : ≥ 100 mg/mL (267.77 mM)
SMILES O=C(C1=CC=CN=C1NCC2=CC=NC=C2)NC3=CC(NCC4(C)C)=C4C=C3
Peso molecular (g/mol) 373.45
Formula Weight (peso de la fórmula) 373.45
Porcentaje de pureza 98.08%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease