Learn More
Tocris Bioscience™ Montelukast sodium
Descripción
Montelukast sodium is a cysteinyl leukotriene receptor 1 (CysLT1) and GPR17 antagonist. Blocks pentylenetetrazole -induced seizures, blood brain barrier (BBB) disruption and leukocyte migration in mice. Reduces neuroinflammation, improves BBB integrity, enhances learning and memory, and restores hippocampal neurogenesis and cognitive function in aged rats. Orally active and brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic acid sodium salt |
| CAS | 151767-02-1 |
| Cantidad | 10 mg |
| Objetivo | Leukotriene and Related Receptor Antagonists |
| Fórmula molecular | C35H35ClNNaO3S |
| Formula Weight (peso de la fórmula) | 608.17 |
| Pureza | >99% |