Learn More
Tocris Bioscience™ Montelukast sodium
CysLT1 and GPR17 antagonist
Marca: Tocris Bioscience™ 5904/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Montelukast sodium is a cysteinyl leukotriene receptor 1 (CysLT1) and GPR17 antagonist. Blocks pentylenetetrazole -induced seizures, blood brain barrier (BBB) disruption and leukocyte migration in mice. Reduces neuroinflammation, improves BBB integrity, enhances learning and memory, and restores hippocampal neurogenesis and cognitive function in aged rats. Orally active and brain penetrant.Especificaciones
| 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic acid sodium salt | |
| 151767-02-1 | |
| 10 mg |