Learn More
Tocris Bioscience™ MOMBA
Descripción
MOMBA is a selective orthosteric agonist for human free fatty acid 2 (hFFA2) engineered receptors, designer receptors activated by designer drugs (DREADD), it shows no activity to wild-type hFFA2, hFFA3, and mouse FFA2. MOMBA inhibit forskolin-stimulated cAMP levels in cells expressing the hFFA2-DREADD receptor with improved potency and equivalent efficacy to Sorbic acid (Cat. No. 7119). In hFFA2-DREADD transgenic mice, MOMBA treatment significantly reduces gut transit and promotes concentration-dependent release of GLP-1 from colonic crypts. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Methoxy-3-methylbenzoic acid |
| CAS | 6880-04-2 |
| Cantidad | 250 mg |
| Objetivo | DREADD Activators |
| Fórmula molecular | C9H10O3 |
| Pureza | 0.98 |