Learn More
Ansell Model 111 - AlphaTec™ 3000 Ultrasonically Welded and Taped Protective Suit
Tienda Ansell ProductosDescripción
- Reinforced protection: The AlphaTec™ 3000 Ultrasonically Welded and Taped - Model 111 protective suit's multi-layered fabric offers a robust barrier against various biohazards and inorganic chemicals
- Enhanced features: This protective barrier is bolstered by the inclusion of dual-zip systems, double cuffs and a double zip enclosure that boosts wear life
- Increased visibility: Designed in a high-visibility bright yellow color, this protective outer garment remains noticeable even in low-lit environments, boosting wearer and workplace safety
- Assured antistatic properties: In addition, it adheres to EN 1149-5 antistatic standards
- Design features: Coverall with 2-piece hood, double zip closure, double cuffs, elasticated hood, waist, double cuffs and ankles.
Especificaciones
Especificaciones
| Tipo de producto | Hooded Coveralls |
| Tamaño | Large |
| Sexo | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Desechables | Single-use |
| Estilo de cintura | Elastic |
| Estilo de tobillo | Elastic |
| Estilo de manguito | Double |
| Estilo de cuello | Hooded |
| Mostrar más |