missing translation for 'onlineSavingsMsg'
Learn More

MNK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Código de producto. 16826323 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16826323 20 μL 20 microlitros
16816323 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16826323 Proveedor Proteintech N.º de proveedor 101361AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

MKNK1 encodes a predicted 424-amino acid protein containing a sequence typical of the catalytic domain of serine/threonine kinases and a putative N-terminal nuclear localization signal sequence. C-terminal region of MKNK1 was phosphorylated and activated in vivo and in vitro by ERK1 and p38 MAP kinases, but not by JNK/SAPK. The activated ERK is essential for the activation of MKNK1 in insect cells. MKNK1 was activated following stimulation of HeLa cells with a variety of extracellular stimuli. MNK1 bind tightly to the growth factor-regulated MAP kinases, Erk1 and Erk2, and also binds strongly to the stress-activated kinase, p38.

Especificaciones

Antígeno MNK1
Aplicaciones Immunocytochemistry, Western Blot, Immunofluorescence
Clasificación Polyclonal
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica MKNK1
N.º de referencia del gen O08605, Q4G050, Q9BUB5
Alias de gen MKNK1, MNK1
Símbolos de los genes Mknk1
Especie del huésped Rabbit
Inmunógeno MNK1 Fusion Protein Ag0184
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17346, 500526, 8569
Especies diana Mouse, Rat, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.