missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP15 Polyclonal Antibody

Descripción
MMP15 Polyclonal Antibody for Western Blot, IHC (P), Flow
MMPs are a group of enzymes involved in matrix degradation. Of the sixteen members of matrix metalloproteinase family ten exist in soluble form whereas MT-MMPs exist as integral membrane proteins. MT1-MMP, MT2-MMP, MT3-MMP also known as MMP14, MMP15, MMP16 respectively contain a C-terminal transmembrane domain anchoring them to the cell surface. They have an 8 amino acid insert in their catalytic domain. MT1-MMP cleaves progelatinase A (MMP-2) and progelatinase B to their active forms. MT2-MMP plays a minor role in activation of pro-MMP-2 to its active form in breast carcinomas. Coexpression of MT1-MMP and MT2-MMp in advanced stages of breast carcinomas is indicative of possible involvement of MT2-MMP in its metastasis.
Especificaciones
Especificaciones
| Antígeno | MMP15 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | Mmp15 |
| N.º de referencia del gen | O54732, P51511 |
| Alias de gen | AI503551; matrix metallo protease; matrix metallopeptidase 15; matrix metallopeptidase 15 (membrane-inserted); matrix metalloproteinase 15; matrix metalloproteinase 15 (membrane-inserted); matrix metalloproteinase-15; Membrane type 2-MMP; membrane-type matrix metalloproteinase 2; Membrane-type-2 matrix metalloproteinase; MMP; Mmp15; MMP-15; MMPs; MT2MMP; MT2-MMP; MT-MMP 2; MTMMP2; SMCP-2 |
| Símbolos de los genes | Mmp15 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?