Learn More
Tocris Bioscience™ MM 102
GREENER_CHOICE
WDR5/MLL interaction inhibitor
Marca:  Tocris Bioscience™ 5307/10
Encompass
Código de producto. 15776675
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
MM 102 is a potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM). Inhibits MLL1 methyltransferase activity and MLL-1-induced HoxA9 and Meis-1 gene expression in leukemia cells expressing the MLL1-AF9 fusion gene. Also inhibits cell growth and induces apoptosis in leukemia cells harbouring MLL1 fusion proteins.
Especificaciones
Especificaciones
>98%
MM 102
MM 102
1883545-52-5
783.83
10 mg
Certificados
Promociones
Promociones
Título del producto
Seleccione un problema
For Research Use Only