Learn More
ML365, MedChemExpress
Código de producto. 16471347
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16471347 50 mg 50 miligramos
16455836 100 mg 100 miligramos
16437837 5 mg 5 miligramos
16405118 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16471347 Proveedor Medchem Express N.º de proveedor HY12345
MedChemExpress ML365 is a selective two-pore domain potassium channel KCNK3/TASK1 inhibitor, with an IC50 of 4 nM. ML365 acts as a pharmacological tool that can be used to examine the specific roles of TASK1 channels.
ML365 is a selective two-pore domain potassium channel KCNK3/TASK1 inhibitor, with an IC50 of 4 nM. ML365 acts as a pharmacological tool that can be used to examine the specific roles of TASK1 channels.
Especificaciones
Nombre del producto químico o material ML365
CAS 947914-18-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C22H20N2O3
Cantidad 50 mg
Información de solubilidad DMSO : ≥ 100 mg/mL (277.46 mM)
SMILES O=C(C1=C(OC)C=CC=C1)NC2=CC=CC(NC(C3=CC=CC(C)=C3)=O)=C2
Peso molecular (g/mol) 360.41
Formula Weight (peso de la fórmula) 360.41
Porcentaje de pureza 98.91%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation