Learn More
ML364, MedChemExpress
Product Code. 16454486
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16454486 100 mg 100 miligramos
16445947 5 mg 5 miligramos
16423218 10 mg 10 miligramos
16419766 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16454486 Supplier Medchem Express Supplier No. HY100900
This item is not returnable. View return policy
MedChemExpress ML364 is a selective ubiquitin specific peptidase 2 (USP2) inhibitor (IC50=1.1 μM) with anti-proliferative activity, which direct binds to USP2 (Kd=5.2 μM), induces an increase in cellular cyclin D1 degradation and causes cell cycle arrest. ML364 increases the levels of mitochondrial ROS and decreases in the intracellular content of ATP.
ML364 is a selective ubiquitin specific peptidase 2 (USP2) inhibitor (IC50=1.1 μM) with anti-proliferative activity, which direct binds to USP2 (Kd=5.2 μM), induces an increase in cellular cyclin D1 degradation and causes cell cycle arrest. ML364 increases the levels of mitochondrial ROS and decreases in the intracellular content of ATP.
Specifications
Nombre del producto químico o material ML364
CAS 1991986-30-1
Color off White
Notas de grado de pureza Research
Fórmula molecular C24H18F3N3O3S2
Cantidad 100 mg
Información de solubilidad DMSO : ≥ 33 mg/mL (63.76 mM)
SMILES O=C(NC1=NC(C2=CC=CC=C2)=CS1)C3=CC=C(C(F)(F)F)C=C3NS(=O)(C4=CC=C(C)C=C4)=O
Peso molecular (g/mol) 517.54
Formula Weight (peso de la fórmula) 517.54
Porcentaje de pureza 99.94%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death