Learn More
Tocris Bioscience™ ML 382
MRGPRX1 positive allosteric modulator
Marca: Tocris Bioscience™ 5738/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ML 382 is a selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.Especificaciones
| Mas-Related G Protein Coupled Receptor Modulators | |
| 1646499-97-9 | |
| 50 mg |