Learn More
Tocris Bioscience™ ML 315 hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Marca: Tocris Bioscience™ 5700/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ML 315 hydrochloride is a inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.Especificaciones
| Inhibitor of Clk and DYRK Kinases | |
| 1440251-53-5 | |
| 410.68g/mol | |
| Cdc2-like Kinase Inhibitors |
| Cdc2-like Kinase | |
| C18H13Cl2N3O2.HCl | |
| Store at -20°C | |
| 50 mg |
Título del producto
For Research Use Only