Learn More
Tocris Bioscience™ ML 299
Descripción
ML 299 is a selective allosteric modulator and potent inhibitor of phospholipases D1 and D2 (IC50 values are 6 and 12 nM, respectively). ML 299 decreases invasive migration in glioblastoma cells and increases caspase 3/7 activity in serum-free conditions. ML 299 is non-cytotoxic, active in vivo and CNS penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Bromo-N-[(1 S)-2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5 ]dec-8-yl]-1-methylethyl]benzamide |
| CAS | 1426916-00-8 |
| Cantidad | 2 mg |
| Objetivo | Phospholipase Inhibitors |
| Fórmula molecular | C23H26BrFN4O2 |
| Pureza | 0.98 |