Learn More
Tocris Bioscience™ (±)-ML 209
RORγt inverse agonist; suppresses Th17 cell differentiation
Marca: Tocris Bioscience™ 5987/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
(±)-ML 209 is a RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).Especificaciones
| White | |
| Nuclear Receptor Antagonists | |
| C25H31NO6 | |
| Soluble to 100mM in DMSO | |
| Inverse agonism Th17 cell differentiation, inhibits MS multiple sclerosis |
| Small Molecule Inhibitor | |
| 1334526-14-5 | |
| 441g/mol | |
| Store at -20°C | |
| 10 mg |
Título del producto
For Laboratory Research Use Only