Learn More
MKT-077, MedChemExpress
Código de producto. 16416318
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16416318 10 mg 10 miligramos
16468897 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16416318 Proveedor Medchem Express N.º de proveedor HY1509610mg
MedChemExpress MKT-077 is a rhodacyanine dye and also a heat shock protein 70 (Hsp70) inhibitor which exhibits significant antitumor activity.
MKT-077 is a rhodacyanine dye and also a heat shock protein 70 (Hsp70) inhibitor which exhibits significant antitumor activity.
Especificaciones
Nombre del producto químico o material MKT-077
CAS 147366-41-4
Notas de grado de pureza Research
Fórmula molecular C21H22ClN3OS2
Sinónimo FJ-776
Información de solubilidad DMSO : 56.67 mg/mL (131.18 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES CN(C1=CC=CC=C1S/2)C2=C3C(N(CC)/C(S\3)=C/C4=[N+](CC)C=CC=C4)=O.[Cl-]
Peso molecular (g/mol) 432.0
Formula Weight (peso de la fórmula) 432.0
Porcentaje de pureza 98.0%
Grado Research
Color Marrón
Cantidad 10 mg
Forma física Sólido