Learn More
MK-571 sodium, MedChemExpress
Código de producto. 16493927
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
16493927 50 mg 50 miligramos
16493726 25 mg 25 miligramos
16460468 5 mg 5 miligramos
16428008 10 mg 10 miligramos
4 options
Código de producto. 16493927
Marca: Medchem Express HY19989A
MedChemExpress MK-571 (L-660711) sodium is an orally active, potent and selective competitive leukotriene D4 (LTD4) receptor antagonist, with Ki values of 0.22 and 2.1 nM in guinea pig and human lung membranes, respectively. MK-571 sodium is also a inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1). MK-571 sodium inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
MK-571 (L-660711) sodium is an orally active, potent and selective competitive leukotriene D4 (LTD4) receptor antagonist, with Ki values of 0.22 and 2.1 nM in guinea pig and human lung membranes, respectively. MK-571 sodium is also a inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1). MK-571 sodium inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
Especificaciones
Nombre del producto químico o material MK-571 sodium
Forma física Solid
CAS 115103-85-0
Color Yellow
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C26H26ClN2NaO3S2
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo L-660711 sodium
Información de solubilidad H2O : 33.33 mg/mL (62.06 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C(O[Na])CCSC(C1=CC=CC(/C=C/C2=NC3=CC(Cl)=CC=C3C=C2)=C1)SCCC(N(C)C)=O
Peso molecular (g/mol) 537.07
Formula Weight (peso de la fórmula) 537.07
Porcentaje de pureza 98.0%