Learn More
SP Genevac™ miVac™ Replacement lid seal
Descripción
- Designed as a direct replacement lid seal for miVac concentrators to maintain optimal sealing performance
- Compatible with both miVac Duo and Quattro Concentrator systems for versatile use
- Helps ensure proper vacuum conditions for efficient sample concentration
- Manufactured for reliable fit and long-lasting performance in laboratory environments
Especificaciones
Especificaciones
| Material | EPDM |
| Tipo | Seal |
| Para utilizar con (equipo) | Quattro Concentrator |