Learn More
Mitiglinide calcium hydrate, MedChemExpress
Código de producto. 16462196
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
500 mg
Tamaño de la unidad:
100 miligramos
500 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16462196 500 mg 500 miligramos
16438416 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16462196 Proveedor Medchem Express N.º de proveedor HYB0682A
MedChemExpress Mitiglinide calcium hydrate (KAD-1229), an insulinotropic agent, is an ATP-sensitive K+ (KATP) channel antagonist. Mitiglinide calcium hydrate is highly specific to the Kir6.2/SUR1 complex (the pancreatic beta-cell KATP channel). Mitiglinide Calcium hydrate can be used for the research of type 2 diabetes.
Mitiglinide calcium hydrate (KAD-1229), an insulinotropic agent, is an ATP-sensitive K+ (KATP) channel antagonist. Mitiglinide calcium hydrate is highly specific to the Kir6.2/SUR1 complex (the pancreatic beta-cell KATP channel). Mitiglinide Calcium hydrate can be used for the research of type 2 diabetes.
Especificaciones
Nombre del producto químico o material Mitiglinide calcium hydrate
Forma física Solid
CAS 207844-01-7
Color Blanco
Cantidad 500 mg
Notas de grado de pureza Research
Fórmula molecular C19H24NO3.1/2Ca.H2O
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism
Sinónimo KAD-1229 S-21403
Información de solubilidad DMSO : 25 mg/mL (70.93 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N1C[C@@](CCCC2)([H])[C@@]2([H])C1)C[C@@H](C([O-])=O)CC3=CC=CC=C3.[0.5Ca2+].O
Peso molecular (g/mol) 352.46
Formula Weight (peso de la fórmula) 352.46
Porcentaje de pureza 99.9%