Learn More
Greiner Bio-One MiniCollect™ Tube, Gold Cap, CAT Serum Sep Clot Activator, Non-Sterile
Descripción
- The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
- Serum separator tubes contain a separator gel in the base of the tube; during centrifugation, this gel forms a stable barrier between the serum and the blood cells.
Especificaciones
Especificaciones
| Aditivo | CAT Serum Sep Clot Activator/Gel Separator |
| Esterilidad | No estéril |
| Capacidad (métrico) | 0.5/0.8 mL |
| Cierre incluido | Yes |
| Color del tapón | Gold |
| Cantidad | 500/paq. |