Learn More
Aceite mineral, Thermo Scientific Chemicals
Código de producto. 11318307
Cambiar vista
Click to view available options
Cantidad:
1 kg
5 x 1 kg
Tamaño de la unidad:
1 Kg
5 Kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11318307 5 x 1 kg 5 Kg
11328307 1 kg 1 Kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Normativa: Este producto está sujeto a un impuesto especial sobre los hidrocarburos de acuerdo con la Directiva Europea 2003/96/CE.Lamentamos informarle que este producto no está disponible para Canarias, Ceuta y Melilla. Disculpe las molestias.
Código de producto. 11318307 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 031911.D9
Solicitar formato a granel o personalizado
Normativa: Este producto está sujeto a un impuesto especial sobre los hidrocarburos de acuerdo con la Directiva Europea 2003/96/CE.Lamentamos informarle que este producto no está disponible para Canarias, Ceuta y Melilla. Disculpe las molestias.
CAS: 8020-83-5
Mineral oil is used as an overlay covering microdrops of culture medium in petri dishes. It is an active ingredient in baby lotions, cold creams, ointments and cosmetics. It is also used in electrical and mechanical industries as thermal fluid in electrical components and non -conductive coolant. It finds application as a transformer oil, as an anti rust agent for blades and as an insulator in high voltage switchgear. Further, it is used to store reactive metals like lithium, sodium, etc and in X-ray crystallography to suspend crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 8020-83-5
Número MDL MFCD00131611
Especificaciones
Nombre del producto químico o material Mineral oil
Color Incolora
Temperatura de inflamación >100°C (212°F)
Olor Mineral-oil-like
Cantidad 5 x 1 kg
Información de solubilidad Miscible with ether. Slightly miscible with ethanol. Immiscible with water.
Forma física Líquido