Learn More
Tocris Bioscience™ Miglustat hydrochloride
Descripción
Miglustat hydrochloride is an orally active α-glucosidase I and II and ceramide-specific glycosyltransferase inhibitor. Rescues trafficking-deficient F508del-CFTR in human airway epithelial cells via inhibition of ER α-glucosidases I and II. Also has broad spectrum antiviral activity.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Miglustat hydrochloride |
| CAS | 210110-90-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C10H22ClNO4 |
| Formula Weight (peso de la fórmula) | 255.74 |
| Pureza | >98% |
| Peso molecular | 255.739 |