missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MIF ELISA Kit

Código de producto. p-7231032
Cambiar vista
Click to view available options
Cantidad:
5 × 96 pruebas
96 pruebas
Tamaño de la unidad:
1 set
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
15405073 96 pruebas 1 set
17135509 5 × 96 pruebas cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15405073 Proveedor Invitrogen™ N.º de proveedor EHMIF
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

ELISA

The Human Macrophage Inflammatory Factor (Hu MIF) ELISA quantitates Hu MIF in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MIF. Principle of the method The Human MIF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

MIF (macrophage migration inhibitory factor) was one of the first cytokine activities to be discovered and was initially described as a T cell-derived factor that inhibits the random migration of macrophages (Weiser 1989). Recently, MIF was rediscovered as a pituitary hormone that act as the counterregulatory hormone for glucocorticoid action within the immune system (Bernhagen 1993, Mitchell 1995). MIF was released from macrophages and T cells in response to physiological concentrations of glucocorticoids. The secreted MIF counter-regulates the immunosuppressive effects of steroids on immune cell activation and cytokine production (Bucala 1998). MIF plays a critical role in the host control of inflammation and immunity.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P14174
Intervalo del ensayo 8,23-6000 pg/ml
Sensibilidad del ensayo 6 pg
Intervalo de calibración De 6 pg/mL a 6000 pg/ml
Contenido y almacenamiento Placas prerecubiertas, patrón o calibrador, anticuerpo detector, conjugado de HRP, diluyentes, tampón de lavado, sustratos y solución de parada. Almacene el kit a 2-8 °C. Consulte el manual del producto para conocer el contenido detallado y las condiciones de almacenamiento para obtener la máxima estabilidad.
ensayo ELISA sandwich de fase sólida
Conjugado Biotina
Formato del kit de inmunoensayo Sandwich ELISA Kit
Tipo de producto Kit de ELISA
Familia de proteínas Citocinas y receptores
Tipo de muestra Plasma, Sobrenadante, Suero
Especificidad Este kit de ELISA no muestra reactividad cruzada con ninguna de las citocinas analizadas: Angiogenina humana, BDNF, BLC, ENA-78, FGF-4, IL-1 alfa, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, leptina, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alfa, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alfa, TNF-beta, TPO, VEGF.
Suficiente para 96 ensayos
Especies diana Humano
Para utilizar con (aplicación) ELISA, ensayos de proteínas, biología de proteínas, biología del cáncer, inmunología, neurociencia, neurología, detección de proteínas, análisis de proteínas
Para utilizar con (equipo) Lector de microplacas colorimétrico
Alias de gen cytokine; delayed early response protein 6; DER6; GIF; GLIF; glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; HGNC:7097; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migration inhibitory factor; macrophage migration inhibitory factor (glycosylation-inhibiting factor); MGC107654; Mif; MMIF; phenylpyruvate tautomerasecytokine; delayed early response protein 6; DER6; GIF; GLIF; glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; HGNC:7097; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migration inhibitory factor; macrophage migration inhibitory factor (glycosylation-inhibiting factor); MGC107654; Mif; MMIF; phenylpyruvate tautomerase;
ID de gen (Entrez) 4282
Símbolo de gen MIF
CV interensayo < 12 %
CV intraensayo < 10 %
Contenido del kit Placa de 96 pocillos recubiertos previamente, Estándar, Concentrado de diluyente de ensayo, Anticuerpo de detección biotinilado, SAV-HRP, Tampón de lavado, Cromógeno, Solución de parada, Cubiertas adhesivas para placas
Etiqueta o tinte Peroxidasa de rábano picante (HRP)
Cantidad 96 pruebas
Estado normativo RUO
Disciplina de investigación Inmunología
Volumen de muestra 50 μl de plasma; 50 μl de suero; 100 μl de sobrenadante
Condiciones de envío Wet or Dry Ice
Requisitos de almacenamiento De 2 °C a 8 °C
Sinónimo GIF, GLIF, MMIF
Objetivo Humano
Tiempo de prueba 1 h 20 min
Tiempo total del ensayo 4 h 45 min
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.