Learn More
Micronomicin sulfate, 590 ^mg/mg, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar J66072.09
| Cantidad | 10g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- An aminoglycoside antibiotic commonly used against gram negative bacteria
Especificaciones
| Micronomicin sulfate | |
| 66803-19-8 | |
| 10g | |
| C20H43N5O11S | |
| 667°C | |
| MQDGQSCLOYLSEK-SCFBDNQUSA-N | |
| (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(methylamino)methyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid | |
| 171808 |
| 590μg/mg | |
| White to Yellow | |
| >110°C (230°F) | |
| MFCD00941451 | |
| Freely soluble in water | |
| OS(O)(=O)=O.CNC[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1 | |
| 561.65 | |
| 561.65 |