Learn More
Microbiologics™ EZ-CFU™ One-Step Microorganism Preparations
Código de producto. 11952745
Cambiar vista
Click to view available options
Especie:
Bacillus subtilis subesp. spizizenii
ATCC®:
6633
Formato:
EZ-CFU
Tamaño de la unidad:
paquete de 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Especie ATCC® Formato Tamaño de la unidad
11952745 Bacillus subtilis subesp. spizizenii 6633 EZ-CFU paquete de 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 11952745 Proveedor Microbiologics™ N.º de proveedor 0486Z
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
missing translation for 'validMainframeMsg'
Includes
Product warranty and a peel-off Certificate of Assay
Lyophilized microorganism preparations deliver less than 100CFU per 0.1mL of hydrated suspension—no dilutions are required
EZ-CFU One-Step preparations are designed to provide the required microorganism challenge concentrations for tests such as Growth Promotion Tests, Microbial Limits Tests and Microbial Enumeration Tests. These ready-to-use, dependable, and cost-effective enumerated microorganism preparations allow laboratories to conveniently and confidently conquer a variety of mandated Pharmacopoeial challenges.
  • Up to 19 tests per 2mL aliquot or 190 total tests per kit can be completed
  • Produces a durable, non-foaming suspension that is ready to be added to the challenge material immediately following the hydration step (30 minutes)
  • For added convenience, a peel-off Certificate of Assay is included for quality control documentation
  • Traceable to reference cultures
Información de órdenes
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Especificaciones
ATCC® 6633
Formato EZ-CFU
Tipo de producto EZ-CFU
Incluye Product warranty and peel-off certificate of assay
Tipo de empaquetado Vial
Especie Bacillus subtilis subesp. spizizenii