Learn More
Tocris Bioscience™ MI 14
Descripción
MI 14 is a potent and selective PI 4-K IIIβ inhibitor (IC50 = 54 nM). Exhibits >1,800-fold selectivity for PI 4-K IIIβ over PI 4-K IIIα and PI 4-K IIα, respectively. Exhibits antiviral activity against single-stranded positive sense RNA viruses in HeLa cell-based assays.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[2-[[6-Chloro-3-[3-[(dimethylamino)sulfonyl]phenyl]-2-methylimidazo[1,2-b]pyridazin-8-yl]amino]ethyl]acetamide |
| CAS | 1715934-43-2 |
| Cantidad | 10 mg |
| Objetivo | Other Kinase Inhibitors |
| Fórmula molecular | C19H23ClN6O3S |
| Formula Weight (peso de la fórmula) | 450.94 |
| Pureza | 98% |