Learn More
Tocris Bioscience™ MI 14
Potent and selective PI 4-kinase IIIβ inhibitor
Marca: Tocris Bioscience™ 5604/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MI 14 is a potent and selective PI 4-K IIIβ inhibitor (IC50 = 54 nM). Exhibits >1,800-fold selectivity for PI 4-K IIIβ over PI 4-K IIIα and PI 4-K IIα, respectively. Exhibits antiviral activity against single-stranded positive sense RNA viruses in HeLa cell-based assays.Especificaciones
| N-[2-[[6-Chloro-3-[3-[(dimethylamino)sulfonyl]phenyl]-2-methylimidazo[1,2-b]pyridazin-8-yl]amino]ethyl]acetamide | |
| 1715934-43-2 | |
| 10 mg |