Learn More
Tocris Bioscience™ Metyrapone
Descripción
Cytochrome P450 inhibitor. Blocks glucocorticoid synthesis via inhibition of steroid 11-β hydroxylase (CYP11B1) activity (IC50 = 7.83 μM). Also inhibits CYP3A4 and cytochrome P450-mediated ω/ω-1 hydroxylase activity.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Metyrapone |
| CAS | 54-36-4 |
| Cantidad | 50 mg |
| Fórmula molecular | C14H14N2O |
| Formula Weight (peso de la fórmula) | 226.27 |
| Pureza | >99% |
| Peso molecular | 226.28 |