missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobencenosulfonato de metilo, 97 %, Thermo Scientific™

Catalog Number 15447318
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1421261-59-7
C7H7BrO3S
251.09
MFCD22683148
RBLPSOBJNBVDMD-UHFFFAOYSA-N
3-Bromobenzenesulfonic acid methyl ester
21222348
3-bromobencenosulfonato de metilo
COS(=O)(=O)C1=CC(Br)=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
1421261-59-7
C7H7BrO3S
251.09
MFCD22683148
RBLPSOBJNBVDMD-UHFFFAOYSA-N
3-Bromobenzenesulfonic acid methyl ester
21222348
3-bromobencenosulfonato de metilo
COS(=O)(=O)C1=CC(Br)=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl 3-bromobenzenesulfonate
Cantidad 250 mg
Formula Weight (peso de la fórmula) 251.09
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'