Learn More
Metil 3,4,5-trihidroxibenzoato, 98 %, Thermo Scientific Chemicals
Código de producto. 11372286
Cambiar vista
Click to view available options
Cantidad:
50 g
250 g
Tamaño de la unidad:
250 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11372286 250 g 250 g
11362286 50 g 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11372286 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor L05926.30
Solicitar formato a granel o personalizado
This item is not returnable. View return policy
CAS: 99-24-1 | C8H8O5 | 184.147 g/mol
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El metil 3,4,5-trihidroxibenzoato muestra una potente actividad antitumoral al inhibir la infiltración del tumor en las células T reguladoras CD4+CD25+.
Solubilidad
Ligeramente soluble en agua.
Notas
Almacenar en un lugar fresco y seco, en un recipiente bien sellado. Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-24-1
Fórmula molecular C8H8O5
Peso molecular (g/mol) 184.147
Número MDL MFCD00002194
Clave InChI FBSFWRHWHYMIOG-UHFFFAOYSA-N
Sinónimo methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
PubChem CID 7428
Nombre IUPAC metil 3,4,5-trihidroxibenzoato
SMILES COC(=O)C1=CC(=C(C(=C1)O)O)O
Specifications
Nombre del producto químico o material Methyl 3,4,5-trihydroxybenzoate
Punto de fusión 198°C to 203°C
Cantidad 250 g
Beilstein 2113180
Índice Merck 144345
Información de solubilidad Slightly soluble in water.
Formula Weight (peso de la fórmula) 184.15
Porcentaje de pureza 0.98