Learn More
Metaxalona-d3 (3,5-dimetilfenoxi-2,4,6-d3), CDN
Product Code. 30320207
Change view
Click to view available options
Cantidad:
0.005 g
0.01 g
Unit Size:
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30320207 0.005 g 0.01 g
30310594 0.01 g 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320207 Supplier CDN Supplier No. CDND64140.005G
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Descubra más de 4000 compuestos deuterados de alta calidad, con envío directo desde stock para mayor rapidez. Ofrecemos muchas API únicas, incluidos ingredientes farmacéuticos activos, nitrosoaminas, aminoácidos, esteroides, gases, pesticidas y ácidos grasos.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Metaxalone-D3 (3,5-dimethylphenoxy-2,4,6-D3)
CAS 1192812-66-0
Fórmula molecular C12 D3 H12 N O3
Cantidad 0.005 g
Sinónimo 2-Oxazolidinone, 5-[(3,5-dimethylphenoxy-2,4,6-d3)methyl]- (ACI), 5-[(3,5-Dimethylphenoxy-2,4,6-d3)methyl]-2-oxazolidinone (ACI), Metaxalone-d3, Metaxalone-D3 (3,5-dimethylphenoxy-2,4,6-D3)
Fórmula InChI InChI=1 S/C12H15NO3/c1-8-3-9(2)5-10(4-8)15-7-11-6-13-12(14)16-11/h3-5,11 H,6-7H2,1-2H3,(H,13,14)/i3D,4 D,5 D
SMILES [2 H]c1c(C)c([2 H])c(OCC2CNC(=O)O2)c([2 H])c1C
Nombre IUPAC 5-[(2,4,6-trideuterio-3,5-dimetilfenoxi)metil]-1,3-oxazolidin-2-ona
Peso molecular (g/mol) 224.2709
Formula Weight (peso de la fórmula) 224.124 g/mol
Porcentaje de pureza 99 atom % D, min 98% Chemical Purity