Learn More
MedChemExpress Melanotan I
Product Code. 16491718
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
Unit Size:
5 miligramos
Product Code. Cantidad unitSize
16491718 5 mg 5 miligramos
1 options
This item is not returnable. View return policy
Product Code. 16491718 Supplier MedChemExpress Supplier No. HYN2466
This item is not returnable. View return policy
MedChemExpress Melanotan I is a potent non-selective melanocortin receptor (MCR) agonist. Melanotan I is a synthetic analogue of α-melanocyte stimulating hormone (α-MSH) that stimulates melanogenesis. Melanotan I can induce skin tanning by mimicking the actions of a-MSH on the melanocortin type 1 receptors (MC1R) of melanocytes. Melanotan I can be used for sunlight-induced skin cancers research.
Specifications
Pureza 97.22%
CAS 75921-69-6
Contenido y almacenamiento Powder -80°C 2 years, -20°C 1 year∣The compound is unstable in solutions, freshly prepared is recommended.
Peso molecular 1646.85 g/mol
Tipo de producto Peptides
Cantidad 5 mg
Condiciones de envío Room temperature
Solubilidad H2O : ≥ 50 mg/mL (30.36 mM)
Sinónimo MT-I∣ [Nle4,D-Phe7]-α-MSH