Learn More
Diatrizoato de meglumina, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | Diatrizoic acid meglumine salt |
| CAS | 131-49-7 |
| Fórmula molecular | C11 H9 I3 N2 O4 . C7 H17 N O5 |
| Cantidad | 5 g |
| Sinónimo | Diatrizoic acid meglumine, D-Glucitol, 1-deoxy-1-(methylamino)-, 3,5-bis(acetylamino)-2,4,6-triiodobenzoate (1:1) (ACI), D-Glucitol, 1-deoxy-1-(methylamino)-, 3,5-bis(acetylamino)-2,4,6-triiodobenzoate (salt) (9CI), Benzoic acid, 3,5-bis(acetylamino)-2,4,6-triiodo-, compd. with 1-deoxy-1-(methylamino)-D-glucitol (1:1) (9CI), Benzoic acid, 3,5-diacetamido-2,4,6-triiodo-, compd. with 1-deoxy-1-(methylamino)glucitol (7CI), Benzoic acid, 3,5-diacetamido-2,4,6-triiodo-, compd. with 1-deoxy-1-methylaminosorbitol (6CI), Glucitol, 1-deoxy-1-(methylamino)-, 3,5-diacetamido-2,4,6-triiodobenzoate (salt), D- (8CI), Sorbitol, 1-deoxy-1-methylamino-, compd. with 3,5-diacetamido-2,4,6-triiodobenzoic acid (6CI), 3,5-Diacetamido-2,4,6-triiodobenzoic acid methylglucamine salt, Amidotrizoic acid methylglucamine salt, Angiografin, Angiografin 310, Angiografin 60, Angiografin 65, Angiographin, Angiovist 282, Cardiografin, Diatrizoate meglumine, Diatrizoate methylglucamine, Ditrizoate methylglucamine, Gastrografin, Hypaque 13.4, Hypaque 60, Hypaque M 30, Hypaque meglumine, Meglumine amidotrizoate, Meglumine diatrizoate, Methylglucamine 3,5-diacetamido-2,4,6-triiodobenzoate, Methylglucamine diacetyldiaminetriiodobenzoate, Methylglucamine diatrixoate, Methylglucamine diatrizoate, Methylglucammonium 2,4,6-triiodo-3,5-dipropionamidobenzoate, N-Methylglucamine diatrizoate, Reno M 60, Reno-M-Dip, Renograffin M 76, Renografin, Renurix, Unipaque, Urografic acid methylglucamine salt, Urovist |
| Fórmula InChI | InChI=1S/C11H9I3N2O4.C7H17NO5/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18;1-8-2-4(10)6(12)7(13)5(11)3-9/h1-2H3,(H,15,17)(H,16,18)(H,19,20);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1 |
| SMILES | CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC(=O)Nc1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I |
| Nombre IUPAC | ácido 3,5-diacetamido-2,4,6-triyodobenzoico;(2R,3R,4R,5S)-6-(metilamino)hexano-1,2,3,4,5-pentol |
| Peso molecular (g/mol) | 809.13 |
| Formula Weight (peso de la fórmula) | 808.8803 |