Learn More
Medroxiprogesterona, LoGiCal
Specifications
Specifications
| Nombre del producto químico o material | Medroxyprogesterone |
| CAS | 520-85-4 |
| Nombres de analitos o componentes | Medroxyprogesterone |
| Condiciones de envío | Room Temperature |
| Fórmula molecular | C22 H32 O3 |
| Certificaciones/conformidad | ISO 9001 |
| Sinónimo | Medroxyprogesterone Acetate Imp. B (EP), Medroxyprogesterone Acetate USP Related Compound B, Medroxyprogesterone Acetate USP RC B, Medroxyprogesterone, 17-Hydroxy-6alpha-methylpregn-4-ene-3,20-dione, Pregn-4-ene-3,20-dione, 17-hydroxy-6-methyl-, (6alpha)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6alpha-methyl- (7CI,8CI), Progesterone, 17-hydroxy-6alpha-methyl- (6CI), (6alpha)-17-Hydroxy-6-methylpregn-4-ene-3,20-dione, 17-Hydroxy-6alpha-methylprogesterone, 17alpha-Hydroxy-6alpha-methylprogesterone, 6alpha-Methyl-17-hydroxyprogesterone, 6alpha-Methyl-17R-hydroxyprogesterone, 6alpha-Methyl-17alpha-hydroxypregn-4-ene-3,20-dione, 6alpha-Methyl-17alpha-hydroxyprogesterone, 6alpha-Methyl-4-pregnen-17alpha-ol-3,20-dione, Medoxyprogesterone, Medroxyprogesteron, NSC 27408, U 8840, Medroxyprogesterone Acetate Related Compound B, Medroxyprogesterone Acetate Impurity B |
| Fórmula InChI | InChI=1S/C22H32O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h12-13,16-18,25H,5-11H2,1-4H3/t13-,16+,17-,18-,20+,21-,22-/m0/s1 |
| Tipo de solución | Neat |
| SMILES | C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(O)C(=O)C)[C@@]4(C)CCC(=O)C=C14 |
| Show More |