Learn More
Medroxi Progesterona-D6 17-Acetato, TRC
Product Code. 30479705
Change view
Click to view available options
Cantidad:
1 mg
Unit Size:
1 miligramo
Product Code. Cantidad unitSize
30479705 1 mg 1 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30479705 Supplier Toronto Research Chemicals Supplier No. TRCM2035621MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Medroxyprogesterone 17-acetate-d6 (6alpha-methyl-d3,acetyl-d3)
CAS 2376036-01-8
Notas de grado de pureza HPLC
Fórmula molecular C24 2H6 H28 O4
Cantidad 1 mg
Sinónimo Pregn-4-ene-3,20-dione, 17-(acetyl-2,2,2-d3-oxy)-6-(methyl-d3)-, (6α)-, (6α)-17-(Acetyl-2,2,2-d3-oxy)-6-(methyl-d3)-pregn-4-ene-3,20-dione, Medroxy progesterone-d6 17-acetate, Medroxyprogesterone-d6 acetate, Medroxyprogesterone Acetate D6 (6α-methyl-D3,acetyl-D3)
Fórmula InChI InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1/i1D3,3D3
SMILES [2H]C([2H])([2H])[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C([2H])([2H])[2H])C(=O)C)[C@@]4(C)CCC(=O)C=C14
Nombre IUPAC [(6S,8R,9S,10R,13S,14S,17R)-17-acetil-10,13-dimetil-3-oxo-6-(trideuteriometil)-2,6,7,8,9,11,12,14,15,16-decahidro-1H-ciclopenta[a]fenantren-17-il] 2,2,2-trideuterioacetato
Peso molecular (g/mol) 392.56
Formula Weight (peso de la fórmula) 392.2834
Porcentaje de pureza >95