missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED29 Polyclonal Antibody

Descripción
Immunogen sequence: LELCLRLAHEC LSQSCDSAKH SPTLVPTATK PDAVQPDSLP YPQYLAVIKA QISCAKDIHT ALLDCAN Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multi-protein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med29 (mediator complex subunit 29), also known as IXL (intersex-like), is a 200 amino acid nuclear protein and component of the mediator complex. Widely expressed in embryo and adult tissue, Med29 is considered a novel amplification target gene in pancreatic cancer and is encoded by a gene that maps to human chromosome 19q13.2. Chromosome 19 consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.
Especificaciones
Especificaciones
| Antígeno | MED29 |
| Aplicaciones | Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | MED29 |
| N.º de referencia del gen | Q9NX70 |
| Alias de gen | 2810405O22Rik; AU020902; chunp6869; DKFZp434H247-like protein; intersex-like protein; ixl; ixl protein; MED2; MED29; Mediator complex subunit 29; mediator of RNA polymerase II transcription subunit 29; RGD1311009; similar to Homo sapiens DKFZp434H247 protein; zgc:109789 |
| Símbolos de los genes | MED29 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?