missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDC Monoclonal Antibody (D1), Invitrogen™
Descripción
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.
Especificaciones
Especificaciones
| Antígeno | MDC |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | D1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | CCL22 |
| N.º de referencia del gen | O00626 |
| Alias de gen | A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?