missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCM3 Recombinant Rabbit Monoclonal Antibody (JB99-35)
GREENER_CHOICE

Código de producto. 17135373
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17135373 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17135373 Proveedor Invitrogen™ N.º de proveedor MA534701

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse thymus tissue lysate, NIH-3T3, Hela, 293T, HepG2, SH-SY-5Y, rat testis tissue, human tonsil tissue, human placenta tissue, mouse small intestine tissue, K562.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MCM3
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon JB99-35
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
génica MCM3
N.º de referencia del gen P25205
Alias de gen AL033361; C80350; cervical cancer proto-oncogene 5; DNA polymerase alpha holoenzyme-associated protein P1; DNA replication factor MCM3; DNA replication licensing factor MCM3; HCC5; hRlf beta subunit; MCM3; MCM3 minichromosome maintenance deficient 3; Mcmd; Mcmd3; MGC1157; mini chromosome maintenance deficient; minichromosome maintenance complex component 3; minichromosome maintenance deficient 3; minichromosome maintenance deficient 3 (S. cerevisiae); P1; P1.h; p1.m; p102; P1-MCM3; replication licensing factor, beta subunit; RLF subunit beta; RLFB; RP1-108C2.3
Símbolos de los genes MCM3
Especie del huésped Rabbit
Inmunógeno Synthetic peptide within Human MCM3 aa 1-50.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4172
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.