Learn More
MCL1 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
Código de producto. 17592941
Encompass_Preferred
Change view
Click to view available options
Cantidad:
30 μL
100 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17592941 30 μL 30 microlitros
17572941 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17592941 Proveedor Origene Technologies N.º de proveedor TA500982S
Mouse Monoclonal Antibody
MCL1 Monoclonal antibody specifically detects MCL1 in Human samples. It is validated for Immunocytochemistry, Western Blot
Especificaciones
Antígeno MCL1
Aplicaciones Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon OTI2D6
Concentración 0.69 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica Srr
N.º de referencia del gen Q9GZT4
Alias de gen AW556805; BCL2 family apoptosis regulator; BCL2L3; bcl2-L-3; Bcl-2-like protein 3; bcl-2-related protein EAT/mcl1; EAT; Induced myeloid leukemia cell differentiation protein Mcl-1; induced myeloid leukemia cell differentiation protein Mcl-1 homolog; Mcl1; Mcl-1; mcl1/EAT; MCL1-ES; MCL1L; MCL1S; MCL-1S; MGC104264; MGC1839; myeloid cell leukemia 1; myeloid cell leukemia ES; myeloid cell leukemia sequence 1; myeloid cell leukemia sequence 1 (BCL2-related); TM
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human SRR produced in HEK293T cell.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 63826
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1